Decyl aldehyde, 95%, Thermo Scientific Chemicals
Decyl aldehyde, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Decyl aldehyde, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number 154975000
Price (USD)/ Each
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namedecanal
Molecular FormulaC10H20O
InChI KeyKSMVZQYAVGTKIV-UHFFFAOYSA-N
SMILESCCCCCCCCCC=O
Molecular Weight (g/mol)156.27
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=94 %
Infrared spectrumConforms
Refractive index1.4270 to 1.4320 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only