Thermo Scientific Chemicals

2'-Deoxyadenosine hydrate, 99%

Catalog number: 154990250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2'-Deoxyadenosine hydrate, 99%

Catalog number: 154990250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 154990250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16373-93-6
IUPAC Name
(2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Formula
C10H13N5O3
InChI Key
OLXZPDWKRNYJJZ-YUZWJPFSNA-N
SMILES
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
Appearance (Color)
White to light beige
Appearance (Form)
Powder or crystalline powder
Infrared spectrum
Conforms
HPLC
>=98.5 %
Water
=<8 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text