Chemical Identifiers
IUPAC Name2-{[(1R,2R)-2-[bis(carboxylatomethyl)azaniumyl]cyclohexyl](carboxylatomethyl)azaniumyl}acetate
Molecular FormulaC14H20N2O8
InChI KeyFCKYPQBAHLOOJQ-NXEZZACHSA-L
SMILES[O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol)344.32
View more