Thermo Scientific Chemicals

2,3-Diaminopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 155020100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2,3-Diaminopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 155020100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 155020100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
452-58-4
IUPAC Name
pyridine-2,3-diamine
Molecular Formula
C5H7N3
InChI Key
ZZYXNRREDYWPLN-UHFFFAOYSA-N
SMILES
NC1=CC=CN=C1N
Appearance (Form)
Crystalline powder or needles
Infrared spectrum
Conforms
Appearance (Color)
Grey to dark brown
Appearance
May darken on storage
Titration with HClO4
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text