Thermo Scientific Chemicals

1,3-Dibromopropane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 155041000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

1,3-Dibromopropane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 155041000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 155041000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-64-8
IUPAC Name
1,3-dibromopropane
Molecular Formula
C3H6Br2
InChI Key
VEFLKXRACNJHOV-UHFFFAOYSA-N
SMILES
BrCCCBr
GC
>=97.5 %
Appearance (Color)
Clear colorless to yellow
Refractive index
1.5220 to 1.5240 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text