1x1 image pixel for data collection
UltraPure™ Tris Hydrochloride
UltraPure™ Tris Hydrochloride
Invitrogen™

UltraPure™ Tris Hydrochloride

UltraPure™ Tris Hydrochloride [tris(hydroxymethyl)aminomethane hydrochloride] is used to simplify preparation of Tris buffers.Read more
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Catalog NumberQuantity
15506017Promo Image500 g
Catalog number 15506017
Price (USD)
155.65
Online Exclusive
169.00
Save 13.35 (8%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
155.65
Online Exclusive
169.00
Save 13.35 (8%)
Each
Add to cart
UltraPure™ Tris Hydrochloride [tris(hydroxymethyl)aminomethane hydrochloride] is used to simplify preparation of Tris buffers.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialTris
For Use With (Application)Nucleic Acid Gel Electrophoresis, Blotting
FormPowder
Product LineUltraPure™
Product TypeTris Hydrochloride
Quantity500 g
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Unit SizeEach
Contents & Storage
Store at room temperature.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link