Thermo Scientific Chemicals

Diethyl methylmalonate, 99%, Thermo Scientific Chemicals

Catalog number: 155141000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl methylmalonate, 99%, Thermo Scientific Chemicals

Catalog number: 155141000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 155141000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
609-08-5
IUPAC Name
1,3-diethyl 2-methylpropanedioate
Molecular Formula
C8H14O4
InChI Key
UPQZOUHVTJNGFK-UHFFFAOYSA-N
SMILES
CCOC(=O)C(C)C(=O)OCC
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4120 to 1.4140 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text