Thermo Scientific Chemicals

2,4-Dihydroxybenzophenone 99%, Thermo Scientific Chemicals

Catalog number: 155181000
Each
Thermo Scientific Chemicals

2,4-Dihydroxybenzophenone 99%, Thermo Scientific Chemicals

Catalog number: 155181000
Each
Catalog number: 155181000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
131-56-6
IUPAC Name
4-benzoylbenzene-1,3-diol
Molecular Formula
C13H10O3
InChI Key
ZXDDPOHVAMWLBH-UHFFFAOYSA-N
SMILES
OC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
Loss on drying
=<0.1 %
Infrared spectrum
Conforms
Melting point
144°C to 147°C
Appearance (Form)
Crystalline powder
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text