UltraPure™ TEMED
UltraPure™ TEMED
Invitrogen™

UltraPure™ TEMED

UltraPure™ TEMED (N,N,N´,N´-tetramethylethylenediamine) is the most commonly used accelerator for the polymerization of acrylamide gels.Read more
Have Questions?
Catalog NumberQuantity
1552401030 mL
Catalog number 15524010
Price (USD)
79.50
Each
Add to cart
Quantity:
30 mL
Request bulk or custom format
Price (USD)
79.50
Each
Add to cart
UltraPure™ TEMED (N,N,N´,N´-tetramethylethylenediamine) is the most commonly used accelerator for the polymerization of acrylamide gels.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Gel Electrophoresis
Product LineUltraPure™
Product TypeTEMED
Quantity30 mL
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C, dark.

Frequently asked questions (FAQs)

Do you carry ammonium persulfate and TEMED?

Yes, details below:

- Ammonium Persulfate: Cat. No. 17874
- TEMED: Cat. No. 15524010 and Cat. No. 17919

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link