Thermo Scientific Chemicals

(3,4-Dimethoxyphenyl)acetonitrile, 99+%, Thermo Scientific Chemicals

Catalog number: 155265000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

(3,4-Dimethoxyphenyl)acetonitrile, 99+%, Thermo Scientific Chemicals

Catalog number: 155265000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 155265000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-17-4
IUPAC Name
2-(3,4-dimethoxyphenyl)acetonitrile
Molecular Formula
C10H11NO2
InChI Key
ASLSUMISAQDOOB-UHFFFAOYSA-N
SMILES
COC1=CC=C(CC#N)C=C1OC
Infrared spectrum
Conforms
GC
>=99.0 %
Appearance (Color)
White to off-white
Sulfated ash
=<0.1 %
Appearance (Form)
Crystals or crystalline needles

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text