1x1 image pixel for data collection
Dimethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Dimethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Dimethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Dimethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethylamine hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 155270050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS506-59-2
IUPAC Namehydrogen dimethylamine chloride
Molecular FormulaC2H8ClN
InChI KeyIQDGSYLLQPDQDV-UHFFFAOYSA-N
SMILES[H+].[Cl-].CNC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
pH5 to 6 (5 % in water)
Appearance (Color)White to almost white
Titration Argentometric98.5 to 101.5 % (on dry substance)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0466069Certificate of AnalysisSep 11, 2024155270050, 155275000, 155270010, 155270025
A0465754Certificate of AnalysisAug 29, 2024155270050, 155275000, 155270010, 155270025
A0460091Certificate of AnalysisFeb 21, 2024155270050, 155275000, 155270010, 155270025
A0455166Certificate of AnalysisJan 05, 2024155270050, 155275000, 155270010, 155270025
A0458403Certificate of AnalysisDec 01, 2023155270050, 155275000, 155270010, 155270025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection