UltraPure™ Glycine
Inquire about OEM or Commercial Supply version of this product here.
UltraPure™ Glycine
Invitrogen™

UltraPure™ Glycine

UltraPure™ Glycine is an amino acid used in the preparation of some electrophoresis buffers. With a pKa1 of 2.3 andRead more
Have Questions?
Catalog number 15527013
Price (USD)/ Each
64.65
Online exclusive
70.50 
Save 5.85 (8%)
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
64.65
Online exclusive
70.50 
Save 5.85 (8%)
Add to cart
UltraPure™ Glycine
Catalog number15527013
Price (USD)/ Each
64.65
Online exclusive
70.50 
Save 5.85 (8%)
-
Add to cart
UltraPure™ Glycine is an amino acid used in the preparation of some electrophoresis buffers. With a pKa1 of 2.3 and a pKa2 of 9.6, UltraPure™ Glycine is suitable for buffering at the extremes of the common buffering range.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialGlycine
Product LineUltraPure™
Product TypeGlycine
Quantity500 g
PurityMolecular Biology Grade
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
FormPowder
Unit SizeEach
Contents & Storage
Store at 15°C to 30°C.