Thermo Scientific Chemicals

Diphenylborinic acid 2-aminoethyl ester, 98%, Thermo Scientific Chemicals

Catalog number: 155420050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Diphenylborinic acid 2-aminoethyl ester, 98%, Thermo Scientific Chemicals

Catalog number: 155420050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 155420050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
524-95-8
IUPAC Name
2-aminoethyl diphenylborinate
Molecular Formula
C14H16BNO
InChI Key
BLZVCIGGICSWIG-UHFFFAOYSA-N
SMILES
NCCOB(C1=CC=CC=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
Melting point
190°C to 194°C
Titration with HClO4
>=97.5 %
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text