Thermo Scientific Chemicals

1-Dodecanol, 98%, Thermo Scientific Chemicals

Catalog number: 155455000
500 mL, Each
Thermo Scientific Chemicals

1-Dodecanol, 98%, Thermo Scientific Chemicals

Catalog number: 155455000
500 mL, Each
Quantity
Catalog number: 155455000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-53-8
IUPAC Name
dodecan-1-ol
Molecular Formula
C12H26O
InChI Key
LQZZUXJYWNFBMV-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCO
Appearance (Color)
Colorless to white
Appearance (Form)
Liquid or crystalline solid
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text