Thermo Scientific Chemicals

n-Heptylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 155460250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

n-Heptylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 155460250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 155460250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-68-2
IUPAC Name
heptan-1-amine
Molecular Formula
C7H17N
InChI Key
WJYIASZWHGOTOU-UHFFFAOYSA-N
SMILES
CCCCCCCN
Appearance (Form)
Liquid
Water
=<0.3 %
Infrared spectrum
Conforms
Refractive index
1.4233 to 1.4253 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text