n-Heptylamine, 99+%, Thermo Scientific Chemicals
n-Heptylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Heptylamine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 155465000
Price (USD)/ Each
328.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
386.00 
Save 57.35 (15%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
328.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
386.00 
Save 57.35 (15%)
Add to cart
n-Heptylamine, 99+%, Thermo Scientific Chemicals
Catalog number155465000
Price (USD)/ Each
328.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
386.00 
Save 57.35 (15%)
-
Add to cart
Chemical Identifiers
CAS111-68-2
IUPAC Nameheptan-1-amine
Molecular FormulaC7H17N
InChI KeyWJYIASZWHGOTOU-UHFFFAOYSA-N
SMILESCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.3 %
Infrared spectrumConforms
Refractive index1.4233 to 1.4253 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only