Thermo Scientific Chemicals

Isoamylamine, 99%, Thermo Scientific Chemicals

Catalog number: 155470250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Isoamylamine, 99%, Thermo Scientific Chemicals

Catalog number: 155470250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 155470250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
107-85-7
IUPAC Name
3-methylbutan-1-amine
Molecular Formula
C5H13N
InChI Key
BMFVGAAISNGQNM-UHFFFAOYSA-N
SMILES
CC(C)CCN
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Water
=<0.5 %
Appearance (Form)
Liquid
GC
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text