Thermo Scientific Chemicals

3-Methylbenzylamine, 98%, Thermo Scientific Chemicals

Catalog number: 155480250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Methylbenzylamine, 98%, Thermo Scientific Chemicals

Catalog number: 155480250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 155480250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
100-81-2
IUPAC Name
(3-methylphenyl)methanaminium
Molecular Formula
C8H12N
InChI Key
RGXUCUWVGKLACF-UHFFFAOYSA-O
SMILES
CC1=CC=CC(C[NH3+])=C1
Appearance (Form)
Liquid
Refractive index
1.5340 to 1.5380 (20°C, 589 nm)
GC
>=97.5 %
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text