UltraPure™ 10% SDS Solution is a premixed aqueous solution containing 10% sodium dodecyl sulfate (SDS). SDS is a protein denaturantRead more
Promotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Change view
Catalog Number
Quantity
15553027
4 x 100 mL
24730020
1 L
2 Options
Catalog number 15553027
Price (USD)
102.65
Online Exclusive
112.00
Save 9.35 (8%)
Each
In stock
Add to cart
Quantity:
4 x 100 mL
Request bulk or custom format
Price (USD)
102.65
Online Exclusive
112.00
Save 9.35 (8%)
Each
Add to cart
UltraPure™ 10% SDS Solution is a premixed aqueous solution containing 10% sodium dodecyl sulfate (SDS). SDS is a protein denaturant used in polyacrylamide gel electrophoresis for molecular weight determinations. It also is used to dissociate nucleic acid-protein complexes in DNA extraction protocols, to disrupt cell membranes, and to prepare prehybridization and hybridization solutions. UltraPure™ 10% SDS Solution is filtered and ready to use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Nucleic Acid Gel Electrophoresis, Blotting, DNA Extraction
FormLiquid
Product LineUltraPure™
Product TypeSDS Solution
Quantity4 x 100 mL
Reagent TypeSDS
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.
Frequently asked questions (FAQs)
Why do solutions containing SDS form precipitates?
SDS is a detergent used to denature proteins leading to disruption of cell membranes and dissociation of nucleic acid-protein complexes in DNA and RNA extractions. It tends to come out of solution at concentrations of 10% or greater, especially at low temperatures or in high-salt buffers. SDS can be resolubilized by slowly heating the solution to 65°C with slight agitation.