Thermo Scientific Chemicals

N-Ethylethylenediamine, 98+%, Thermo Scientific Chemicals

Catalog number: 155550500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Ethylethylenediamine, 98+%, Thermo Scientific Chemicals

Catalog number: 155550500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 155550500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
110-72-5
IUPAC Name
(2-azaniumylethyl)(ethyl)azanium
Molecular Formula
C4H14N2
InChI Key
SCZVXVGZMZRGRU-UHFFFAOYSA-P
SMILES
CC[NH2+]CC[NH3+]
GC
>=98.0 %
Appearance (Form)
Liquid
Refractive index
1.4380 to 1.4400 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text