Thermo Scientific Chemicals

N-Methyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals

Catalog number: 155572500
250 mL, Each
Thermo Scientific Chemicals

N-Methyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals

Catalog number: 155572500
250 mL, Each
Quantity
Catalog number: 155572500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6291-84-5
IUPAC Name
(3-aminopropyl)(methyl)amine
Molecular Formula
C4H12N2
InChI Key
QHJABUZHRJTCAR-UHFFFAOYSA-N
SMILES
CNCCCN
Appearance (Form)
Clear liquid
Color scale
=<50 APHA
GC
>=98.5 %
Water
=<0.5 % (K.F.)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text