Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Learn about a safer electrophoresis workflow using the Invitrogen SYBR™ Safe DNA Gel Stain »
Invitrogen™

UltraPure™ TAE Buffer, 10X

Catalog number: 15558026
Have Questions?
Invitrogen™

UltraPure™ TAE Buffer, 10X

Catalog number: 15558026
Have Questions?
Catalog Number
15558026
Unit Size
4 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
155580264 L
-
155580421 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
UltraPure™ 10X TAE Buffer is a sterile-filtered solution of 400 mM Tris-acetate and 10 mM EDTA. TAE Buffer is the most commonly used buffer for agarose DNA electrophoresis. It is supplied in 1 L plastic bottles or in a 4 L or 10 L stackable Cubitainer™ Box. A 1X TAE Buffer solution contains 40 mM Tris-acetate and 1 mM EDTA at pH 8.3.

Performance and Quality Testing: No DNase activity detected.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer
Running Buffer
Concentration
10 X
Format
Bottle
Gel Compatibility
Agarose Gels
For Use With (Application)
Nucleic Acid Gel Electrophoresis, Blotting
Product Line
UltraPure™
Product Type
TAE Buffer
Quantity
4 L
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text