Thermo Scientific Chemicals

2-Methylbenzylamine, 96%, Thermo Scientific Chemicals

Catalog number: 155630250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Methylbenzylamine, 96%, Thermo Scientific Chemicals

Catalog number: 155630250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 155630250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
89-93-0
IUPAC Name
1-(2-methylphenyl)methanamine
Molecular Formula
C8H11N
InChI Key
CJAAPVQEZPAQNI-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1CN
Infrared spectrum
Conforms
Refractive index
1.5415 to 1.5435 (20°C, 589 nm)
Appearance (Form)
Liquid
Titration with HClO4
>=95.0 %
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text