N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%, Thermo Scientific Chemicals
N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 155690250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namediethyl(trimethylsilyl)amine
Molecular FormulaC7H19NSi
InChI KeyJOOMLFKONHCLCJ-UHFFFAOYSA-N
SMILESCCN(CC)[Si](C)(C)C
Molecular Weight (g/mol)145.32
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4103 to 1.4123 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only