Thermo Scientific Chemicals

N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%, Thermo Scientific Chemicals

Catalog number: 155691000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%, Thermo Scientific Chemicals

Catalog number: 155691000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 155691000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
996-50-9
IUPAC Name
diethyl(trimethylsilyl)amine
Molecular Formula
C7H19NSi
InChI Key
JOOMLFKONHCLCJ-UHFFFAOYSA-N
SMILES
CCN(CC)[Si](C)(C)C
Infrared spectrum
Conforms
Refractive index
1.4103 to 1.4123 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to yellow
GC
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text