Aminomethanesulfonic acid, 97%, Thermo Scientific Chemicals
Aminomethanesulfonic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aminomethanesulfonic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
Packaging:
Glass bottle
Catalog number 155745000
Price (USD)/ Each
240.00
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
240.00
Add to cart
Aminomethanesulfonic acid, 97%, Thermo Scientific Chemicals
Catalog number155745000
Price (USD)/ Each
240.00
-
Add to cart
Chemical Identifiers
CAS13881-91-9
IUPAC Nameaminomethanesulfonic acid
Molecular FormulaCH5NO3S
InChI KeyOBESRABRARNZJB-UHFFFAOYSA-N
SMILESNCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with NaOH>=96.0 %
Appearance (Color)White
Infrared spectrumConforms
Melting point180°C to 185°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only