Chemical Identifiers
CAS2946-39-6
IUPAC Name(2R,3R,4S,5R)-2-(6-amino-8-bromo-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular FormulaC10H12BrN5O4
InChI KeyVJUPMOPLUQHMLE-SBTUYZBDNA-N
SMILESNC1=C2N=C(Br)N([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
View more