Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Learn about a safer electrophoresis workflow using the Invitrogen SYBR™ Safe DNA Gel Stain »
Invitrogen™

UltraPure™ TBE Buffer, 10X

Catalog number: 15581028
Have Questions?
Invitrogen™

UltraPure™ TBE Buffer, 10X

Catalog number: 15581028
Have Questions?
Catalog Number
15581028
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1558102810 L
-
155810441 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
UltraPure™ 10X TBE Buffer is a sterile-filtered solution of 1 M Tris, 0.9 M boric acid, and 0.01 M EDTA used to prepare 1X buffer for polyacrylamide and agarose gel electrophoresis. UltraPure™ 10X TBE Buffer is available in a 1 L plastic bottle or in a 10 L Cubitainer™ box.

Performance and quality testing
No DNase, RNase, or protease activity detected.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer
Running Buffer
Concentration
10 X
Format
Bottle
Gel Compatibility
Polyacrylamide Gels, Agarose Gels
For Use With (Application)
Nucleic Acid Gel Electrophoresis, Blotting
Product Line
UltraPure™
Product Type
TBE Buffer
Quantity
10 L
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text