Thermo Scientific Chemicals

Alizarin Complexone Dihydrate, Indicator Grade

Catalog number: 155830010
1 g, Each
Thermo Scientific Chemicals

Alizarin Complexone Dihydrate, Indicator Grade

Catalog number: 155830010
1 g, Each
Quantity
Catalog number: 155830010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
455303-00-1
IUPAC Name
2-[(carboxymethyl)[(3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracen-2-yl)methyl]amino]acetic acid dihydrate
Molecular Formula
C19H19NO10
InChI Key
LLNGUADXYFYGGJ-UHFFFAOYSA-N
SMILES
O.O.OC(=O)CN(CC(O)=O)CC1=CC2=C(C(O)=C1O)C(=O)C1=C(C=CC=C1)C2=O
UV Lambda max
425 to 430 nm (C=10 mg/l, Methanol)
Appearance (Form)
Powder
Water
=<9.5 % (K.F.)
Appearance (Color)
Yellow to orange to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text