Chemical Identifiers
CAS455303-00-1
IUPAC Name2-[(carboxymethyl)[(3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracen-2-yl)methyl]amino]acetic acid dihydrate
Molecular FormulaC19H19NO10
InChI KeyLLNGUADXYFYGGJ-UHFFFAOYSA-N
SMILESO.O.OC(=O)CN(CC(O)=O)CC1=CC2=C(C(O)=C1O)C(=O)C1=C(C=CC=C1)C2=O
View more