Thermo Scientific Chemicals

3-Aminobenzoic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 155855000
500 g, Each
Thermo Scientific Chemicals

3-Aminobenzoic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 155855000
500 g, Each
Quantity
Catalog number: 155855000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
99-05-8
IUPAC Name
3-aminobenzoic acid
Molecular Formula
C7H7NO2
InChI Key
XFDUHJPVQKIXHO-UHFFFAOYSA-N
SMILES
NC1=CC=CC(=C1)C(O)=O
Appearance (Form)
Powder
Melting point
>=172°C
Infrared spectrum
Conforms
HPLC
>=99.0 %
Appearance (Color)
White to beige to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text