trans-Crotonyl chloride, 90%, tech., Thermo Scientific Chemicals
trans-Crotonyl chloride, 90%, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-Crotonyl chloride, 90%, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 155910050
Price (USD)/ Each
22.50
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
22.50
Add to cart
trans-Crotonyl chloride, 90%, tech., Thermo Scientific Chemicals
Catalog number155910050
Price (USD)/ Each
22.50
-
Add to cart
Chemical Identifiers
IUPAC Name(2E)-but-2-enoyl chloride
Molecular FormulaC4H5ClO
InChI KeyRJUIDDKTATZJFE-NSCUHMNNSA-N
SMILESC\C=C\C(Cl)=O
Molecular Weight (g/mol)104.53
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=88 %
Appearance (Color)Clear colorless to brown
Infrared spectrumConforms
Refractive index1.4585 to 1.4615 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only