UltraPure™ SSPE, 20X
Inquire about OEM or Commercial Supply version of this product here.
Product Image
Invitrogen™

UltraPure™ SSPE, 20X

UltraPure 20X SSPE is a prepared buffer concentrate formulated for use in nucleic acid hybridizations and blot transfer applications. SuppliedRead more
Have Questions?
Catalog number 15591050
Price (USD)/ Each
460.00
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
460.00
Add to cart
UltraPure™ SSPE, 20X
Catalog number15591050
Price (USD)/ Each
460.00
-
Add to cart
UltraPure 20X SSPE is a prepared buffer concentrate formulated for use in nucleic acid hybridizations and blot transfer applications. Supplied in 1 L or in 10 x 1 L plastic bottles, sterile-filtered UltraPure 20X SSPE contains 3.0 M NaCl, 0.2 M NaH2PO4, and 0.02 M EDTA at pH 7.4.

Performance and quality testing: no DNase or RNase activity detected

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration20X
Quantity10 x 1 L
FormLiquid
Product LineUltraPure™
Unit SizeEach
Contents & Storage
Store at room temperature.