Thermo Scientific Chemicals

trans-Crotonyl chloride, 90%, tech., Thermo Scientific Chemicals

Catalog number: 155915000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

trans-Crotonyl chloride, 90%, tech., Thermo Scientific Chemicals

Catalog number: 155915000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 155915000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
625-35-4
IUPAC Name
(2E)-but-2-enoyl chloride
Molecular Formula
C4H5ClO
InChI Key
RJUIDDKTATZJFE-NSCUHMNNSA-N
SMILES
C\C=C\C(Cl)=O
Appearance (Color)
Clear colorless to brown
Infrared spectrum
Conforms
Refractive index
1.4585 to 1.4615 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=88 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text