TRIzol™ Reagent
Find Nucleic Acid Purification and Analysis technical tips, troubleshooting help, and resources within our new Nucleic Acid Purification and Analysis Support Center.
TRIzol™ Reagent
Invitrogen™

TRIzol™ Reagent

TRIzol™ Reagent is a complete, ready-to-use reagent for the isolation of high-quality total RNA or the simultaneous isolation of RNA,Read more
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
15596026Promo Image100 mL
15596018Promo Image200 mL
Catalog number 15596026
Price (USD)
254.00
Each
Add to cart
Quantity:
100 mL
Price (USD)
254.00
Each
Add to cart

TRIzol™ Reagent is a complete, ready-to-use reagent for the isolation of high-quality total RNA or the simultaneous isolation of RNA, DNA, and protein from a variety of biological samples. This monophasic solution of phenol and guanidine isothiocyanate is designed to isolate separate fractions of RNA, DNA, and proteins from cell and tissue samples of human, animal, plant, yeast, or bacterial origin, within one hour.

Key features of TRIzol™ Reagent include:
• Permits the isolation of RNA, DNA, and protein from the same sample
• Offers superior lysis capability, even with difficult sample types
• Optimized formulations and protocols for tissues, cells, serum, virus, and bacteria

Reliably purify RNA from multiple sample volumes and sources
TRIzol™ Reagent performs well with small quantities of tissue (50–100 mg) and cells (5 × 106), as well as with large quantities of tissue (≥1 g) and cells (>107), and comes with prototcols for purification from samples of human, animal, plant, or bacterial origin. TRIzol™ Reagent maintains the integrity of the RNA due to highly effective inhibition of RNase activity while disrupting cells and dissolving cell components during sample homogenization. The simplicity of the TRIzol™ Reagent method allows simultaneous processing of a large number of samples. The entire procedure can be completed in 1 hour. Total RNA isolated by TRIzol™ Reagent is free of protein and DNA contamination.

Formulated for isolation of multiple molecular targets
TRIzol™ Reagent allows you to perform sequential precipitation of RNA, DNA, and proteins from a single sample. After homogenizing the sample with TRIzol™ Reagent, chloroform is added, and the homogenate is allowed to separate into a clear upper aqueous layer (containing RNA), and interphase and red lower organic layers (containing the DNA and proteins). RNA is precipitated from the aqueous layer with isopropanol. DNA is precipitated from the interphase/organic layer with ethanol. Protein is precipitated from the phenol-ethanol supernatant by isopropanol precipitation. The precipitated RNA, DNA, or protein is washed to remove impurities, and then resuspended for use in downstream applications.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume20 to 600 μL
Final Product TypeTotal RNA, DNA, Protein
For Use With (Application)RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern/Western blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization, enzymatic assays, immunoprecipitation, gel shift assays, 2D gel electrophoresis
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Purification Time1 hr.
Quantity100 mL
Shipping ConditionRoom Temperature
Starting Material AmountBacteria: ≤107 cells
Cells: ≤107
Plant: ≤107 cells, 100 mg
Tissue: ≤100 mg
Yeast: ≤107 cells
YieldDNA: ≤7 μg
RNA: ≤73 μg
Protein: varies
Isolation TechnologyOrganic Extraction
Sample TypeBacteria, Cells, Plants, Tissue, Yeast
Unit SizeEach
Contents & Storage
100 mL TRIzol Reagent; room temperature

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link