5-Amino-1,3,4-thiadiazole-2-thiol, 98%, Thermo Scientific Chemicals
5-Amino-1,3,4-thiadiazole-2-thiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Amino-1,3,4-thiadiazole-2-thiol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 155970500
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.10 
Save 10.45 (14%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.10 
Save 10.45 (14%)
Add to cart
5-Amino-1,3,4-thiadiazole-2-thiol, 98%, Thermo Scientific Chemicals
Catalog number155970500
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.10 
Save 10.45 (14%)
-
Add to cart
Chemical Identifiers
CAS2349-67-9
IUPAC Name5-amino-2,3-dihydro-1,3,4-thiadiazole-2-thione
Molecular FormulaC2H3N3S2
InChI KeyGDGIVSREGUOIJZ-UHFFFAOYSA-N
SMILESNC1=NNC(=S)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow
Appearance (Form)Powder
Titration with NaOH>=97.5 % (On dry substance)
Loss on drying=<1 % (110°C)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only