2-Ethoxyethanol, 99%, extra pure, Thermo Scientific Chemicals
2-Ethoxyethanol, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Ethoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 156020010
Price (USD)/ Each
63.65
Online exclusive
70.20 
Save 6.55 (9%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
63.65
Online exclusive
70.20 
Save 6.55 (9%)
Add to cart
2-Ethoxyethanol, 99%, extra pure, Thermo Scientific Chemicals
Catalog number156020010
Price (USD)/ Each
63.65
Online exclusive
70.20 
Save 6.55 (9%)
-
Add to cart
Chemical Identifiers
CAS110-80-5
IUPAC Name2-ethoxyethan-1-ol
Molecular FormulaC4H10O2
InChI KeyZNQVEEAIQZEUHB-UHFFFAOYSA-N
SMILESCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Specific gravity(20°C) 0.928 to 0.932
Refractive index1.4060 to 1.4100 (20°C, 589 nm)
Color scale=<10 APHA
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only