Thermo Scientific Chemicals

2-Ethoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 156020250
25 L, Metal drum, Each
Thermo Scientific Chemicals

2-Ethoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 156020250
25 L, Metal drum, Each
Quantity
Packaging
Catalog number: 156020250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
110-80-5
IUPAC Name
2-ethoxyethan-1-ol
Molecular Formula
C4H10O2
InChI Key
ZNQVEEAIQZEUHB-UHFFFAOYSA-N
SMILES
CCOCCO
Refractive index
1.4060 to 1.4100 (20°C, 589 nm)
Appearance (Form)
Clear liquid
Specific gravity
(20°C) 0.928 to 0.932
Color scale
=<10 APHA
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text