Thermo Scientific Chemicals

Ethyl chrysanthemate, 95%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 156060250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl chrysanthemate, 95%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 156060250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 156060250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-41-6
IUPAC Name
ethyl 2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylate
Molecular Formula
C12H20O2
InChI Key
VIMXTGUGWLAOFZ-UHFFFAOYNA-N
SMILES
CCOC(=O)C1C(C=C(C)C)C1(C)C
Infrared spectrum
Conforms
GC
>=94 %
Water
=<0.2 %
Refractive index
1.4590 to 1.4610 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text