1500 Series B2 Bio Safety Cabinets 4ft. and 6 ft.
Catalog number: 1561-MAR
Have Questions?
1500 Series B2 Bio Safety Cabinets 4ft. and 6 ft.
Catalog number: 1561-MAR
Have Questions?
1500 Series B2 BSC, NSF listed Class II, Type B2 BSC available in nominal widths of 4 ft (1.2 m) and 6 ft (1.8m). Regional voltages selectable. Incorporates SmartFlow intelligent compensation for downflow, energy efficient DC motor, stainless steel interior and line-of-sight graphical display and user controls for easy access.
Catalog Number
1561-MAR
Unit Size
Each
Description
1500 Series Class II, Type B2 BSC, 6 ft (1.8 m) nominal width with UV, 120 V, 50/60 Hz
Voltage
115 VAC
Price (USD)
Full specifications
Description1500 Series Class II, Type B2 BSC, 6 ft (1.8 m) nominal width with UV, 120 V, 50/60 Hz
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1841 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level66 dBA
Power Consumption370 W
Receptacles2 Duplex, GFI
Width (English) Exterior78 in.
Volume (Metric) Exhaust1696 m3/hr. (with DIM), 2149 m3/hr. (with Traverse)
Width (Metric) Exterior1981 mm
Width (English) Interior72.5 in.
Voltage115 VAC
Amperage12 A
Energy Consumption (Operating Set Point)0.37 KW
Unit SizeEach
Catalog Number
Unit Size
Each
Description
1500 Series Class II, Type B2 BSC, 4 ft (1.2 m) nominal width, 120V, 50/60Hz
Voltage
115 VAC
Price (USD)
Full specifications
Description1500 Series Class II, Type B2 BSC, 4 ft (1.2 m) nominal width, 120V, 50/60Hz
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1232 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level65 dBA
Power Consumption280 W
Receptacles2 Duplex, GFI
Width (English) Exterior54.3 in.
Volume (Metric) Exhaust1130 m3/hr. (with DIM), 1448 m3/hr. (with Traverse)
Width (Metric) Exterior1379 mm
Width (English) Interior48.5 in.
Voltage115 VAC
Amperage12 A
Energy Consumption (Operating Set Point)0.28 KW
Unit SizeEach
Catalog Number
Unit Size
Each
Description
1500 Series Class II, Type B2 BSC, 4 ft (1.2 m) nominal width with UV, 120V, 50/60Hz
Voltage
115 VAC
Price (USD)
Full specifications
Description1500 Series Class II, Type B2 BSC, 4 ft (1.2 m) nominal width with UV, 120V, 50/60Hz
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1232 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level65 dBA
Power Consumption280 W
Receptacles2 Duplex, GFI
Width (English) Exterior54.3 in.
Volume (Metric) Exhaust1130 m3/hr. (with DIM), 1448 m3/hr. (with Traverse)
Width (Metric) Exterior1379 mm
Width (English) Interior48.5 in.
Voltage115 VAC
Amperage12 A
Energy Consumption (Operating Set Point)0.28 KW
Unit SizeEach
Catalog Number
Unit Size
Each
Description
1500 Series Class II, Type B2 BSC, 6 ft (1.8 m) nominal width, 120 V, 50/60 Hz
Voltage
115 VAC
Price (USD)
Full specifications
Description1500 Series Class II, Type B2 BSC, 6 ft (1.8 m) nominal width, 120 V, 50/60 Hz
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1841 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level66 dBA
Power Consumption370 W
Receptacles2 Duplex, GFI
Width (English) Exterior78 in.
Volume (Metric) Exhaust1696 m3/hr. (with DIM), 2149 m3/hr. (with Traverse)
Width (Metric) Exterior1981 mm
Width (English) Interior72.5 in.
Voltage115 VAC
Amperage12 A
Energy Consumption (Operating Set Point)0.37 KW
Unit SizeEach
Catalog Number
Unit Size
Each
Description
1500 Series Class II, Type B2 BSC, 6 ft (1.2m) nominal width with UV, 230 VAC, 50/60 Hz with Type M (India) power cord and internal receptacle
Voltage
230 VAC
Price (USD)
Full specifications
Description1500 Series Class II, Type B2 BSC, 6 ft (1.2m) nominal width with UV, 230 VAC, 50/60 Hz with Type M (India) power cord and internal receptacle
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1841 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level66 dBA
Power Consumption370 W
Receptacles2 Duplex
Width (English) Exterior78 in.
Volume (Metric) Exhaust1696 m3/hr. (with DIM), 2149 m3/hr. (with Traverse)
Width (Metric) Exterior1981 mm
Width (English) Interior72.5 in.
Voltage230 VAC
Amperage12 A
Energy Consumption (Operating Set Point)0.37 KW
Unit SizeEach
Catalog Number
Unit Size
Each
Description
1500 Series Class II, Type B2 BSC with UV, 6 ft (1.8m) nominal width, 230 VAC, 50/60 Hz with Type M (India) power cord and internal receptacle
Voltage
230 VAC
Price (USD)
Full specifications
Description1500 Series Class II, Type B2 BSC with UV, 6 ft (1.8m) nominal width, 230 VAC, 50/60 Hz with Type M (India) power cord and internal receptacle
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1841 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level66 dBA
Power Consumption370 W
Receptacles2 Single
Width (English) Exterior78 in.
Volume (Metric) Exhaust1696 m3/hr. (with DIM), 2149 m3/hr. (with Traverse)
Width (Metric) Exterior1981 mm
Height (Metric)1844 mm (Exterior), 653 mm to 742 mm (Interior)
Width (English) Interior72.5 in.
Height (English) Exterior72.6 in.
Height (English) Interior25.7 in. to 29.2 in.
Voltage230 VAC
Amperage6 A
Energy Consumption (Operating Set Point)0.37 kw
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeDescriptionVoltagePrice (USD)
1561-MARFull specifications
Each1500 Series Class II, Type B2 BSC, 6 ft (1.8 m) nominal width with UV, 120 V, 50/60 Hz115 VACRequest A Quote
Description1500 Series Class II, Type B2 BSC, 6 ft (1.8 m) nominal width with UV, 120 V, 50/60 Hz
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1841 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level66 dBA
Power Consumption370 W
Receptacles2 Duplex, GFI
Width (English) Exterior78 in.
Volume (Metric) Exhaust1696 m3/hr. (with DIM), 2149 m3/hr. (with Traverse)
Width (Metric) Exterior1981 mm
Width (English) Interior72.5 in.
Voltage115 VAC
Amperage12 A
Energy Consumption (Operating Set Point)0.37 KW
Unit SizeEach
1510-MARFull specifications
Each1500 Series Class II, Type B2 BSC, 4 ft (1.2 m) nominal width, 120V, 50/60Hz115 VACRequest A Quote
Description1500 Series Class II, Type B2 BSC, 4 ft (1.2 m) nominal width, 120V, 50/60Hz
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1232 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level65 dBA
Power Consumption280 W
Receptacles2 Duplex, GFI
Width (English) Exterior54.3 in.
Volume (Metric) Exhaust1130 m3/hr. (with DIM), 1448 m3/hr. (with Traverse)
Width (Metric) Exterior1379 mm
Width (English) Interior48.5 in.
Voltage115 VAC
Amperage12 A
Energy Consumption (Operating Set Point)0.28 KW
Unit SizeEach
1511-MARFull specifications
Each1500 Series Class II, Type B2 BSC, 4 ft (1.2 m) nominal width with UV, 120V, 50/60Hz115 VACRequest A Quote
Description1500 Series Class II, Type B2 BSC, 4 ft (1.2 m) nominal width with UV, 120V, 50/60Hz
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1232 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level65 dBA
Power Consumption280 W
Receptacles2 Duplex, GFI
Width (English) Exterior54.3 in.
Volume (Metric) Exhaust1130 m3/hr. (with DIM), 1448 m3/hr. (with Traverse)
Width (Metric) Exterior1379 mm
Width (English) Interior48.5 in.
Voltage115 VAC
Amperage12 A
Energy Consumption (Operating Set Point)0.28 KW
Unit SizeEach
1560-MARFull specifications
Each1500 Series Class II, Type B2 BSC, 6 ft (1.8 m) nominal width, 120 V, 50/60 Hz115 VACRequest A Quote
Description1500 Series Class II, Type B2 BSC, 6 ft (1.8 m) nominal width, 120 V, 50/60 Hz
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1841 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level66 dBA
Power Consumption370 W
Receptacles2 Duplex, GFI
Width (English) Exterior78 in.
Volume (Metric) Exhaust1696 m3/hr. (with DIM), 2149 m3/hr. (with Traverse)
Width (Metric) Exterior1981 mm
Width (English) Interior72.5 in.
Voltage115 VAC
Amperage12 A
Energy Consumption (Operating Set Point)0.37 KW
Unit SizeEach
1565-MARFull specifications
Each1500 Series Class II, Type B2 BSC, 6 ft (1.2m) nominal width with UV, 230 VAC, 50/60 Hz with Type M (India) power cord and internal receptacle230 VACRequest A Quote
Description1500 Series Class II, Type B2 BSC, 6 ft (1.2m) nominal width with UV, 230 VAC, 50/60 Hz with Type M (India) power cord and internal receptacle
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1841 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level66 dBA
Power Consumption370 W
Receptacles2 Duplex
Width (English) Exterior78 in.
Volume (Metric) Exhaust1696 m3/hr. (with DIM), 2149 m3/hr. (with Traverse)
Width (Metric) Exterior1981 mm
Width (English) Interior72.5 in.
Voltage230 VAC
Amperage12 A
Energy Consumption (Operating Set Point)0.37 KW
Unit SizeEach
1565M-MARFull specifications
Each1500 Series Class II, Type B2 BSC with UV, 6 ft (1.8m) nominal width, 230 VAC, 50/60 Hz with Type M (India) power cord and internal receptacle230 VACRequest A Quote
Description1500 Series Class II, Type B2 BSC with UV, 6 ft (1.8m) nominal width, 230 VAC, 50/60 Hz with Type M (India) power cord and internal receptacle
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1841 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level66 dBA
Power Consumption370 W
Receptacles2 Single
Width (English) Exterior78 in.
Volume (Metric) Exhaust1696 m3/hr. (with DIM), 2149 m3/hr. (with Traverse)
Width (Metric) Exterior1981 mm
Height (Metric)1844 mm (Exterior), 653 mm to 742 mm (Interior)
Width (English) Interior72.5 in.
Height (English) Exterior72.6 in.
Height (English) Interior25.7 in. to 29.2 in.
Voltage230 VAC
Amperage6 A
Energy Consumption (Operating Set Point)0.37 kw
Unit SizeEach
Showing 6 of 6
  • NSF listed Class II type B2 BSC
  • SmartFlow intelligent compensation for downflow
  • Energy efficient DC motor
  • Stainless steel interior
  • Line-of-sight graphical display and user controls for easy access
  • Previous models: 1300 Series B2