Thermo Scientific Chemicals

N-Ethylmaleimide, 98+%, Thermo Scientific Chemicals

Catalog number: 156100100
10 g, Each
Thermo Scientific Chemicals

N-Ethylmaleimide, 98+%, Thermo Scientific Chemicals

Catalog number: 156100100
10 g, Each
Quantity
Catalog number: 156100100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
128-53-0
IUPAC Name
1-ethyl-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular Formula
C6H7NO2
InChI Key
HDFGOPSGAURCEO-UHFFFAOYSA-N
SMILES
CCN1C(=O)C=CC1=O
Solubility
(5% in 100% EtOH) Clear to light turbid,
Solubility
Colorless to light yellow solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text