Thermo Scientific Chemicals

4-Fluorobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 156161000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Fluorobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 156161000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 156161000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
456-22-4
IUPAC Name
4-fluorobenzoic acid
Molecular Formula
C7H5FO2
InChI Key
BBYDXOIZLAWGSL-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(F)C=C1
Appearance (Form)
Crystalline powder
Melting point
184°C to 187°C
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text