Thermo Scientific Chemicals

L(+)-Glutamic acid, 99%

Catalog number: 156211000
100 g, Plastic Bottle, Each
Thermo Scientific Chemicals

L(+)-Glutamic acid, 99%

Catalog number: 156211000
100 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 156211000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-86-0
IUPAC Name
2-aminopentanedioic acid
Molecular Formula
C5H9NO4
InChI Key
WHUUTDBJXJRKMK-UHFFFAOYNA-N
SMILES
NC(CCC(O)=O)C(O)=O
Appearance (Color)
White
Appearance (Form)
Powder or crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
98.5 to 100.5 % (On dried substance)
Loss on drying
=<0.5 % (105°C, 3 hrs)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text