2-Hexanol, 99%, Thermo Scientific Chemicals
2-Hexanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Hexanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Packaging:
Glass bottle
Catalog number 156290500
Price (USD)/ Each
96.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
113.00 
Save 16.35 (14%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
96.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
113.00 
Save 16.35 (14%)
Add to cart
2-Hexanol, 99%, Thermo Scientific Chemicals
Catalog number156290500
Price (USD)/ Each
96.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
113.00 
Save 16.35 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namehexan-2-ol
Molecular FormulaC6H14O
InChI KeyQNVRIHYSUZMSGM-UHFFFAOYNA-N
SMILESCCCCC(C)O
Molecular Weight (g/mol)102.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=98.5 %
Refractive index1.4125 to 1.4155 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only