2-Hydroxyethyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
2-Hydroxyethyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Hydroxyethyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 156330025
Price (USD)/ Each
177.65
Online exclusive
197.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
177.65
Online exclusive
197.00 
Save 19.35 (10%)
Add to cart
2-Hydroxyethyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Catalog number156330025
Price (USD)/ Each
177.65
Online exclusive
197.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS868-77-9
IUPAC Name2-hydroxyethyl 2-methylprop-2-enoate
Molecular FormulaC6H10O3
InChI KeyWOBHKFSMXKNTIM-UHFFFAOYSA-N
SMILESCC(=C)C(=O)OCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Acid value=<2 mg KOH/g
Infrared spectrumConforms
Water=<0.2 % (K.F.)
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only