UltraPure™ Calf Thymus DNA Solution
UltraPure™ Calf Thymus DNA Solution
Invitrogen™

UltraPure™ Calf Thymus DNA Solution

UltraPure™ DNA solutions from Calf Thymus, Herring Sperm, or Salmon Sperm are ready-to-use, sheared DNA solutions that are used directlyRead more
Have Questions?
Catalog NumberQuantity
156330195 x 1 mL
Catalog number 15633019
Price (USD)
299.65
Online Exclusive
328.00
Save 28.35 (9%)
Each
-
Add to cart
Quantity:
5 x 1 mL
Request bulk or custom format
Price (USD)
299.65
Online Exclusive
328.00
Save 28.35 (9%)
Each
Add to cart
Ask our AI about this Product

UltraPure™ DNA solutions from Calf Thymus, Herring Sperm, or Salmon Sperm are ready-to-use, sheared DNA solutions that are used directly in the preparation of prehybridization and hybridization solutions. These prepared solutions are used to block the non-specific attachment of probe to the surface of a membrane. The DNA solutions are prepared from highly pure, phenol/chloroform-extracted DNA and DNase-free and RNase-free (DEPC-treated) distilled, deionized water and are sheared to an average size of ≤2,000 bp. The concentration is adjusted to 10 mg/ml.

Performance and Quality Testing: Concentration, size verification, viscosity, and ultraviolet absorbance (A260/A280) are analytically tested.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Chromatin Biology, Nucleic Acid Gel Electrophoresis, Blotting
Key FunctionsBlock Non-Specific Hybridization
Product LineUltraPure™
Product TypeCalf Thymus DNA Solution
Quantity5 x 1 mL
Reagent TypeCalf Thymus DNA
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store at -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link