Thermo Scientific Chemicals

2-Hydroxyethyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 156332500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Hydroxyethyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 156332500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 156332500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
868-77-9
IUPAC Name
2-hydroxyethyl 2-methylprop-2-enoate
Molecular Formula
C6H10O3
InChI Key
WOBHKFSMXKNTIM-UHFFFAOYSA-N
SMILES
CC(=C)C(=O)OCCO
GC
>=96.0 %
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4510 to 1.4530 (20°C, 589 nm)
Stabilizer
180 to 220 ppm MEHQ
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text