4-Hydroxy-3-methoxycinnamic acid, 99%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxycinnamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-3-methoxycinnamic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 156361000
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
Add to cart
4-Hydroxy-3-methoxycinnamic acid, 99%, Thermo Scientific Chemicals
Catalog number156361000
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1135-24-6
IUPAC Name(2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
Molecular FormulaC10H10O4
InChI KeyKSEBMYQBYZTDHS-HWKANZROSA-N
SMILESCOC1=CC(\C=C\C(O)=O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point169°C to 173°C
Appearance (Color)Off-white to beige
Appearance (Form)Powder
Titration with NaOH>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only