Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface
Thermo Scientific™

Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface

Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.
Promo ImagePromotion
Promo code:RCELL51
Stock your lab with plasticware essentials so that you can culture with confidence
From now until Dec 31, get 51% off various cell culture plasticware like dishes, flasks, plates and tubes that help achieve reproducible resultsLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCulture AreaClosure Type
156376Promo Image75 cm²Filter
156372Promo Image25 cm²Filter
156374Promo Image25 cm²Solid
156378Promo Image75 cm²Solid
156380Promo Image175 cm²Filter
156382Promo Image175 cm²Solid
Catalog number 156376
Price (USD)
658.00
Case of 100
Add to cart
Culture Area:
75 cm²
Closure Type:
Filter
Price (USD)
658.00
Case of 100
Add to cart
Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc EasYFlask flasks with Nunclon Supra surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

For Laboratory Use
Specifications
Closure TypeFilter
DescriptionNunc EasYFlask, 75 cm²
No. per Case100 Flasks
No. per Pack5 Flasks
Culture Area75 cm²
Unit SizeCase of 100
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link