Thermo Scientific Chemicals

7-Hydroxy-4-methylcoumarin, 97%

Catalog number: 156385000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

7-Hydroxy-4-methylcoumarin, 97%

Catalog number: 156385000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 156385000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-33-5
IUPAC Name
7-hydroxy-4-methyl-2H-chromen-2-one
Molecular Formula
C10H8O3
InChI Key
HSHNITRMYYLLCV-UHFFFAOYSA-N
SMILES
CC1=CC(=O)OC2=CC(O)=CC=C12
Titration with NaOH
96.0 to 104.0 % (On dry substance)
Water
=<2.5 % (K.F.)
Appearance (Color)
White to light yellow to light beige
Melting point
186°C to 191°C
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text